siRNA Details
| Gene Symbol: Slc25a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053515.1 | NP_445967.1 | 4 | 912 | ||
| GenBank mRNA | D12770.1 | - | 4 | 879 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a4| Gene Symbol: Slc25a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053515.1 | NP_445967.1 | 4 | 912 | ||
| GenBank mRNA | D12770.1 | - | 4 | 879 | ||