siRNA Details
| Gene Symbol: Slc25a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053515.1 | NP_445967.1 | 1 | 199 | ||
| GenBank mRNA | BC060533.1 | - | 1 | 203 | 6922056 | |
| D12770.1 | - | 1 | 166 | |||
| FQ213027.1 | - | 1 | 196 | |||
| FQ213086.1 | - | 1 | 196 | |||
| FQ213612.1 | - | 1 | 225 | |||
| FQ214384.1 | - | 1 | 195 | |||
| FQ215448.1 | - | 1 | 199 | |||
| FQ217254.1 | - | 1 | 195 | |||
| FQ220525.1 | - | 1 | 194 | |||
| FQ224814.1 | - | 1 | 196 | |||

Slc25a4
Genomic Map