siRNA Details
| Gene Symbol: Ngb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033359.3 | NP_203523.2 | 3, 4 | 731 | ||
| GenBank mRNA | AB056655.1 | - | 3, 4 | 303 | ||
| AB056656.1 | - | 2, 3 | 193 | |||
| AF333245.1 | - | 3, 4 | 322 | |||
| AY066001.1 | - | 3, 4 | 531 | |||

Ngb
Genomic Map