siRNA Details
| Gene Symbol: Shc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164060.1 | NP_001157532.1 | 9, 10 | 1457 | ||
| NM_053517.2 | NP_445969.2 | 10, 11 | 1024 | |||
| GenBank mRNA | BC088298.1 | - | 10, 11 | 1024 | 7323045 | |
| CB813924.1 | - | 3, 4 | 305 | |||

Shc1
Genomic Map