siRNA Details
| Gene Symbol: Lyst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053518.1 | NP_445970.1 | 8 | 4198 | ||
| GenBank mRNA | AB020019.1 | - | 8 | 4198 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lyst| Gene Symbol: Lyst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053518.1 | NP_445970.1 | 8 | 4198 | ||
| GenBank mRNA | AB020019.1 | - | 8 | 4198 | ||