siRNA Details
| Gene Symbol: Tdrd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138871.1 | NP_620226.1 | 16 | 3166 | ||
| XM_006238107.2 | XP_006238169.1 | 16 | 2997 | |||
| XM_006238108.2 | XP_006238170.1 | 15 | 2930 | |||
| XM_006238111.2 | XP_006238173.1 | 15 | 3167 | |||
| XM_008763704.1 | XP_008761926.1 | 15 | 3324 | |||
| GenBank mRNA | AB030644.1 | - | 16 | 3166 | ||
| BC090066.1 | - | 15 | 2885 | 7122915 | ||

Tdrd7
Genomic Map