siRNA Details
| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053523.1 | NP_445975.1 | 4 | 423 | ||
| XM_006255133.2 | XP_006255195.1 | 4 | 736 | |||
| GenBank mRNA | AB033771.1 | - | 4 | 423 | ||
| BC061849.1 | - | 4 | 445 | 5621877 | ||

Herpud1
Genomic Map