siRNA Details
| Gene Symbol: Nox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053524.1 | NP_445976.1 | 14 | 1366 | ||
| XM_008759640.1 | XP_008757862.1 | 14 | 1284 | |||
| XM_008759641.1 | XP_008757863.1 | 14 | 1283 | |||
| XM_008759642.1 | XP_008757864.1 | 13 | 1256 | |||
| XM_008759643.1 | XP_008757865.1 | 16 | 1624 | |||
| GenBank mRNA | AB044086.1 | - | 14 | 1334 | ||
| AY027527.1 | - | 14 | 1366 | |||

Nox4
Genomic Map