siRNA Details
| Gene Symbol: Ddx52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053525.1 | NP_445977.1 | 8 | 976 | ||
| GenBank mRNA | AB055628.1 | - | 8 | 976 | ||
| BC089107.1 | - | 8 | 978 | 7310368 | ||
| EF121975.1 | - | 8 | 970 | |||
| EF121976.1 | - | 8 | 970 | |||

Ddx52
Genomic Map