siRNA Details
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138873.2 | NP_620228.1 | 7, 8 | 940 | ||
GenBank mRNA | AF218575.1 | - | 7, 8 | 944 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138873.2 | NP_620228.1 | 7, 8 | 940 | ||
GenBank mRNA | AF218575.1 | - | 7, 8 | 944 |