siRNA Details
| Gene Symbol: Ciita | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270803.1 | NP_001257732.1 | 15, 16 | 3083 | ||
| NM_001270804.1 | NP_001257733.1 | 15, 16 | 3011 | |||
| NM_053529.2 | NP_445981.2 | 15, 16 | 3302 | |||
| XM_006245738.2 | XP_006245800.1 | 15, 16 | 3344 | |||
| XM_008767501.1 | XP_008765723.1 | 16, 17 | 4384 | |||
| GenBank mRNA | AF251305.1 | - | 15, 16 | 3302 | ||
| AF251306.1 | - | 15, 16 | 3083 | |||
| AF251307.1 | - | 15, 16 | 3011 | |||

Ciita
Genomic Map