siRNA Details
| Gene Symbol: Vamp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053531.1 | NP_445983.1 | 1, 2 | 151 | ||
| XM_006249016.2 | XP_006249078.1 | 2, 3 | 304 | |||
| GenBank mRNA | AF281632.1 | - | 1, 2 | 151 | ||
| FQ228923.1 | - | 2, 3 | 264 | |||
| FQ230649.1 | - | 1, 2 | 75 | |||

Vamp7
Genomic Map