siRNA Details
| Gene Symbol: Tpra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053534.2 | NP_445986.2 | 5 | 499 | ||
| XM_006236866.2 | XP_006236928.1 | 4 | 1738 | |||
| XM_006236868.2 | XP_006236930.1 | 4 | 539 | |||
| GenBank mRNA | AF292116.1 | - | 3 | 305 | ||
| FQ228747.1 | - | 5 | 574 | |||

Tpra1
Genomic Map