siRNA Details
| Gene Symbol: Enpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053535.1 | NP_445987.1 | 9 | 874 | ||
| XM_006227692.2 | XP_006227754.1 | 9 | 896 | |||
| GenBank mRNA | AF320054.1 | - | 9 | 874 | ||
| AF340185.1 | - | 9 | 874 | |||
| AF340186.1 | - | 9 | 874 | |||

Enpp1
Genomic Map