siRNA Details
| Gene Symbol: Laptm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053538.2 | NP_445990.2 | 6 | 599 | ||
| GenBank mRNA | AB046592.1 | - | 6 | 611 | ||
| BC060517.1 | - | 6 | 599 | 5597893 | ||
| FQ225663.1 | - | 6 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Laptm5| Gene Symbol: Laptm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053538.2 | NP_445990.2 | 6 | 599 | ||
| GenBank mRNA | AB046592.1 | - | 6 | 611 | ||
| BC060517.1 | - | 6 | 599 | 5597893 | ||
| FQ225663.1 | - | 6 | 597 | |||