siRNA Details
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053539.2 | NP_445991.2 | 5 | 999 | ||
| GenBank mRNA | AF003835.1 | - | 5 | 980 | ||
| BC089786.1 | - | 5 | 999 | 7322086 | ||
| FQ219635.1 | - | 5 | 648 | |||
| FQ220448.1 | - | 5 | 646 | |||
| FQ221966.1 | - | 1 | 48 | |||
| FQ225062.1 | - | 5 | 646 | |||
| FQ226929.1 | - | 5 | 652 | |||
| FQ227874.1 | - | 5 | 649 | |||

Idi1
Genomic Map