siRNA Details
| Gene Symbol: Idi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053539.2 | NP_445991.2 | 5 | 921 | ||
| GenBank mRNA | AF003835.1 | - | 5 | 902 | ||
| BC089786.1 | - | 5 | 921 | 7322086 | ||
| FQ219635.1 | - | 5 | 570 | |||
| FQ220448.1 | - | 5 | 568 | |||
| FQ225062.1 | - | 5 | 568 | |||
| FQ226929.1 | - | 5 | 574 | |||
| FQ227874.1 | - | 5 | 571 | |||

Idi1
Genomic Map