siRNA Details
| Gene Symbol: Lrp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053541.1 | NP_445993.1 | 6 | 1977 | ||
| XM_008759197.1 | XP_008757419.1 | 5 | 3257 | |||
| XM_008759198.1 | XP_008757420.1 | 5 | 3257 | |||
| XM_008759199.1 | XP_008757421.1 | 5 | 3257 | |||
| XM_008759200.1 | XP_008757422.1 | 6 | 2176 | |||
| XM_008759201.1 | XP_008757423.1 | 5 | 3136 | |||
| XM_008759202.1 | XP_008757424.1 | 6 | 2056 | |||
| GenBank mRNA | AB009463.1 | - | 6 | 1977 | ||
| FQ227632.1 | - | 2 | 204 | |||

Lrp3
Genomic Map