siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053544.1 | NP_445996.1 | 3 | 664 | ||
| GenBank mRNA | AF012891.1 | - | 3 | 664 | ||
| AF140346.1 | - | 3 | 815 | |||
| AF220608.1 | - | 912 | ||||
| FQ229224.1 | - | 2 | 95 | |||
More Information
Set Membership: |
|

Sfrp4
Genomic Map