siRNA Details
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105737.1 | NP_001099207.1 | 6 | 1131 | ||
XM_008763910.1 | XP_008762132.1 | 6 | 951 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105737.1 | NP_001099207.1 | 6 | 1131 | ||
XM_008763910.1 | XP_008762132.1 | 6 | 951 |