siRNA Details
| Gene Symbol: Tek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105737.1 | NP_001099207.1 | 22, 23 | 3616 | ||
| XM_008763910.1 | XP_008762132.1 | 22, 23 | 3436 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tek| Gene Symbol: Tek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105737.1 | NP_001099207.1 | 22, 23 | 3616 | ||
| XM_008763910.1 | XP_008762132.1 | 22, 23 | 3436 | |||