siRNA Details
| Gene Symbol: Tie1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053545.1 | NP_445997.1 | 14 | 2433 | ||
| XM_006238720.2 | XP_006238782.1 | 14 | 2495 | |||
| GenBank mRNA | AF030377.1 | - | 2 | 280 | ||
| BC085911.1 | - | 13 | 2631 | |||
| BC169020.1 | - | 14 | 2433 | 9099086 | ||

Tie1
Genomic Map