siRNA Details
| Gene Symbol: Tie1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053545.1 | NP_445997.1 | 2 | 247 | ||
| XM_006238720.2 | XP_006238782.1 | 2 | 315 | |||
| GenBank mRNA | BC085911.1 | - | 2 | 251 | ||
| BC169020.1 | - | 2 | 247 | 9099086 | ||

Tie1
Genomic Map