siRNA Details
| Gene Symbol: Angpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053546.1 | NP_445998.1 | 3 | 766 | ||
| XM_006241609.2 | XP_006241671.1 | 3 | 1108 | |||
| XM_008765463.1 | XP_008763685.1 | 3 | 292 | |||
| XM_008765464.1 | XP_008763686.1 | 3 | 2756 | |||
| GenBank mRNA | AB080023.1 | - | 3 | 766 | ||
| AF030376.1 | - | 3 | 205 | |||
| AF311727.1 | - | 3 | 205 | |||
| AY052399.1 | - | 2 | 183 | |||

Angpt1
Genomic Map