siRNA Details
Gene Symbol: Cx3cl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134455.1 | NP_604450.1 | 3 | 817 | ||
XM_008772329.1 | XP_008770551.1 | 4 | 1554 | |||
GenBank mRNA | AF030358.2 | - | 3 | 817 | ||
BC070938.1 | - | 3 | 818 | 7099250 | ||
Y16813.1 | - | 1 | 188 |
