siRNA Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053549.1 | NP_446001.1 | 3, 4 | 562 | ||
| XM_006230910.2 | XP_006230972.1 | 3, 4 | 659 | |||
| XM_006230911.2 | XP_006230973.1 | 3, 4 | 660 | |||
| XM_006230912.2 | XP_006230974.1 | 3, 4 | 650 | |||
| GenBank mRNA | AF022952.1 | - | 1, 2 | 215 | ||
| AF032925.1 | - | 1, 2 | 158 | |||
| BC076395.1 | - | 3, 4 | 562 | 7109119 | ||

Vegfb
Genomic Map