siRNA Details
| Gene Symbol: Pdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053551.1 | NP_446003.1 | 8 | 900 | ||
| GenBank mRNA | AF034577.1 | - | 8 | 900 | ||
| FQ216954.1 | - | 943 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdk4| Gene Symbol: Pdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053551.1 | NP_446003.1 | 8 | 900 | ||
| GenBank mRNA | AF034577.1 | - | 8 | 900 | ||
| FQ216954.1 | - | 943 | ||||