siRNA Details
| Gene Symbol: Syngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053553.1 | NP_446005.1 | 2 | 344 | ||
| GenBank mRNA | AF039085.1 | - | 2 | 344 | ||
| FQ218897.1 | - | 187 | ||||
| FQ224907.1 | - | 188 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syngr2| Gene Symbol: Syngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053553.1 | NP_446005.1 | 2 | 344 | ||
| GenBank mRNA | AF039085.1 | - | 2 | 344 | ||
| FQ218897.1 | - | 187 | ||||
| FQ224907.1 | - | 188 | ||||