siRNA Details
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001057.2 | NP_001048.2 | 1 | 858 | ||
GenBank mRNA | AK291996.1 | - | 1 | 858 | ||
AK300649.1 | - | 1 | 752 | |||
AY322545.1 | - | 1 | 263 | |||
BC096842.1 | - | 1 | 269 | 6971906 | ||
FJ200479.1 | - | 2 | 155 | |||
M57414.1 | - | 1 | 263 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
