siRNA Details
| Gene Symbol: Bud31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053556.1 | NP_446008.1 | 4 | 446 | ||
| XM_006248887.1 | XP_006248949.1 | 4 | 563 | |||
| XM_006248889.1 | XP_006248951.1 | 5 | 431 | |||
| XM_008769003.1 | XP_008767225.1 | 5 | 660 | |||
| GenBank mRNA | AF058791.1 | - | 4 | 446 | ||
| BC058456.1 | - | 5 | 473 | 6887068 | ||
| FQ230665.1 | - | 2 | 174 | |||
| FQ232789.1 | - | 2 | 174 | |||
| FQ232851.1 | - | 2 | 174 | |||
| FQ232911.1 | - | 2 | 174 | |||
| FQ235300.1 | - | 2 | 174 | |||

Bud31
Genomic Map