siRNA Details
| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053557.1 | NP_446009.1 | 13, 14 | 1427 | ||
| GenBank mRNA | AF059530.1 | - | 13, 14 | 1427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt3| Gene Symbol: Prmt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053557.1 | NP_446009.1 | 13, 14 | 1427 | ||
| GenBank mRNA | AF059530.1 | - | 13, 14 | 1427 | ||