siRNA Details
| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053561.2 | NP_446013.2 | 12 | 1157 | ||
| XM_006241342.1 | XP_006241404.1 | 12 | 1171 | |||
| XM_006241343.2 | XP_006241405.1 | 12 | 1171 | |||
| XM_006241344.2 | XP_006241406.1 | 10 | 908 | |||
| GenBank mRNA | AF062594.1 | - | 12 | 1139 | ||
| CF110129.1 | - | 7 | 592 | |||
| FQ230040.1 | - | 3 | 210 | |||

Nap1l1
Genomic Map