siRNA Details
| Gene Symbol: Nap1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053561.2 | NP_446013.2 | 8 | 755 | ||
| XM_006241342.1 | XP_006241404.1 | 8 | 769 | |||
| XM_006241343.2 | XP_006241405.1 | 8 | 769 | |||
| XM_006241344.2 | XP_006241406.1 | 6 | 506 | |||
| GenBank mRNA | AF062594.1 | - | 8 | 737 | ||
| CF110129.1 | - | 3 | 190 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001146707, NM_015781, AK007322, AK008261, AK012250, and others.

Nap1l1
Genomic Map