siRNA Details
| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053566.1 | NP_446018.1 | 4 | 563 | ||
| GenBank mRNA | AF079162.1 | - | 4 | 805 | ||
| AY357891.1 | - | 4 | 563 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptch1| Gene Symbol: Ptch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053566.1 | NP_446018.1 | 4 | 563 | ||
| GenBank mRNA | AF079162.1 | - | 4 | 805 | ||
| AY357891.1 | - | 4 | 563 | |||