siRNA Details
| Gene Symbol: Cxadr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053570.1 | NP_446022.1 | 4, 5 | 650 | ||
| XM_006248000.2 | XP_006248062.1 | 4, 5 | 751 | |||
| GenBank mRNA | AF109643.1 | - | 4, 5 | 579 | ||
| AF109644.1 | - | 4, 5 | 579 | |||
| BC088313.1 | - | 4, 5 | 650 | 7324723 | ||

Cxadr
Genomic Map