siRNA Details
| Gene Symbol: Sec16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053571.1 | NP_446023.1 | 15 | 1930 | ||
| XM_008769682.1 | XP_008767904.1 | 21 | 5459 | |||
| XM_008769683.1 | XP_008767905.1 | 19 | 5167 | |||
| XM_008769684.1 | XP_008767906.1 | 16 | 2691 | |||
| XM_008769685.1 | XP_008767907.1 | 18 | 4444 | |||
| XM_008769686.1 | XP_008767908.1 | 14 | 1728 | |||
| GenBank mRNA | AB060653.1 | - | 15 | 1930 | ||

Sec16b
Genomic Map