siRNA Details
| Gene Symbol: Prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053576.2 | NP_446028.1 | 2 | 229 | ||
| GenBank mRNA | AF014009.1 | - | 2 | 229 | ||
| FQ218168.1 | - | 2 | 258 | |||
| Y17295.1 | - | 243 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdx6| Gene Symbol: Prdx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053576.2 | NP_446028.1 | 2 | 229 | ||
| GenBank mRNA | AF014009.1 | - | 2 | 229 | ||
| FQ218168.1 | - | 2 | 258 | |||
| Y17295.1 | - | 243 | ||||