siRNA Details
| Gene Symbol: Spp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053577.1 | NP_446029.1 | 239 | |||
| GenBank mRNA | BC088193.1 | - | 239 | 7372900 | ||
| FQ210158.1 | - | 155 | ||||
| FQ210198.1 | - | 176 | ||||
| FQ210350.1 | - | 203 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spp2| Gene Symbol: Spp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053577.1 | NP_446029.1 | 239 | |||
| GenBank mRNA | BC088193.1 | - | 239 | 7372900 | ||
| FQ210158.1 | - | 155 | ||||
| FQ210198.1 | - | 176 | ||||
| FQ210350.1 | - | 203 | ||||