siRNA Details
| Gene Symbol: Spp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053577.1 | NP_446029.1 | 373 | |||
| GenBank mRNA | BC088193.1 | - | 373 | 7372900 | ||
| FQ210158.1 | - | 289 | ||||
| FQ210198.1 | - | 310 | ||||
| FQ210350.1 | - | 337 | ||||
| U19485.1 | - | 154 | ||||

Spp2
Genomic Map