siRNA Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053580.2 | NP_446032.2 | 7 | 1114 | ||
| XM_006252953.2 | XP_006253015.1 | 7 | 1152 | |||
| XM_006252954.1 | XP_006253016.1 | 7 | 1100 | |||
| XM_008771156.1 | XP_008769378.1 | 7 | 1039 | |||
| XM_008771157.1 | XP_008769379.1 | 7 | 1198 | |||
| GenBank mRNA | BC074014.1 | - | 7 | 1114 | 7104661 | |
| U89529.1 | - | 7 | 1011 | |||

Slc27a1
Genomic Map