siRNA Details
| Gene Symbol: Idh3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053581.1 | NP_446033.1 | 11 | 1049 | ||
| XM_006235048.2 | XP_006235110.1 | 11 | 1086 | |||
| GenBank mRNA | AB047540.1 | - | 4 | 318 | ||
| BC079113.1 | - | 11 | 1049 | 7126029 | ||
| FQ214820.1 | - | 11 | 1047 | |||
| FQ215180.1 | - | 11 | 1053 | |||
| FQ215184.1 | - | 11 | 1040 | |||
| FQ215350.1 | - | 11 | 1049 | |||
| FQ217667.1 | - | 5 | 407 | |||

Idh3B
Genomic Map