siRNA Details
| Gene Symbol: Idh3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053581.1 | NP_446033.1 | 5 | 378 | ||
| XM_006235048.2 | XP_006235110.1 | 5 | 415 | |||
| GenBank mRNA | BC079113.1 | - | 5 | 378 | 7126029 | |
| FQ214820.1 | - | 5 | 376 | |||
| FQ215180.1 | - | 5 | 382 | |||
| FQ215184.1 | - | 5 | 370 | |||
| FQ215350.1 | - | 5 | 378 | |||

Idh3B
Genomic Map