siRNA Details
| Gene Symbol: Gosr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053584.2 | NP_446036.1 | 8 | 576 | ||
| XM_006246914.1 | XP_006246976.1 | 8 | 644 | |||
| XM_008768132.1 | XP_008766354.1 | 7 | 447 | |||
| GenBank mRNA | BC126068.1 | - | 8 | 580 | 7387185 | |

Gosr1
Genomic Map