siRNA Details
| Gene Symbol: S100a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053587.1 | NP_446039.1 | 1 | 74 | ||
| XM_006232780.1 | XP_006232842.1 | 2 | 155 | |||
| GenBank mRNA | AB118215.1 | - | 1 | 38 | ||
| L18948.1 | - | 1 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

S100a9| Gene Symbol: S100a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053587.1 | NP_446039.1 | 1 | 74 | ||
| XM_006232780.1 | XP_006232842.1 | 2 | 155 | |||
| GenBank mRNA | AB118215.1 | - | 1 | 38 | ||
| L18948.1 | - | 1 | 74 | |||