siRNA Details
| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053588.2 | NP_446040.1 | 485 | |||
| GenBank mRNA | AF315468.1 | - | 446 | |||
| BC061821.1 | - | 485 | 5599087 | |||
| FQ220857.1 | - | 781 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf138| Gene Symbol: Rnf138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053588.2 | NP_446040.1 | 485 | |||
| GenBank mRNA | AF315468.1 | - | 446 | |||
| BC061821.1 | - | 485 | 5599087 | |||
| FQ220857.1 | - | 781 | ||||