siRNA Details
| Gene Symbol: Ptprr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113390.1 | NP_001106861.1 | 11 | 1401 | ||
| NM_053594.2 | NP_446046.2 | 2110 | ||||
| GenBank mRNA | BC089186.1 | - | 7 | 892 | 7315399 | |
| D38292.1 | - | 2113 | ||||
| D64050.1 | - | 11 | 1429 | |||
| U14914.1 | - | 1457 | ||||

Ptprr
Genomic Map