siRNA Details
| Gene Symbol: Ece1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053596.2 | NP_446048.2 | 17 | 1971 | ||
| XM_006239275.2 | XP_006239337.1 | 17 | 2559 | |||
| XM_006239276.2 | XP_006239338.1 | 16 | 2575 | |||
| GenBank mRNA | BC072504.1 | - | 17 | 1971 | 7099038 | |
| D29683.1 | - | 17 | 2006 | |||

Ece1
Genomic Map