siRNA Details
| Gene Symbol: Nnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270867.1 | NP_001257796.1 | 3 | 1049 | ||
| NM_053601.2 | NP_446053.1 | 3 | 1103 | |||
| NM_181687.2 | NP_859015.1 | 2 | 1022 | |||
| XM_008762390.1 | XP_008760612.1 | 3 | 1093 | |||
| GenBank mRNA | BC127473.1 | - | 2 | 956 | 8364201 | |
| FQ213576.1 | - | 2 | 980 | |||
| U08290.1 | - | 3 | 1020 | |||
| U09785.1 | - | 2 | 939 | |||

Nnat
Genomic Map