siRNA Details
| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053604.2 | NP_446056.1 | 1 | 1400 | ||
| GenBank mRNA | BC081754.1 | - | 1 | 1400 | 7130336 | |
| U21662.1 | - | 1 | 1063 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat2| Gene Symbol: Mgat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053604.2 | NP_446056.1 | 1 | 1400 | ||
| GenBank mRNA | BC081754.1 | - | 1 | 1400 | 7130336 | |
| U21662.1 | - | 1 | 1063 | |||