siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133294.1 | NP_579828.1 | 3 | 1417 | ||
| XM_008763693.1 | XP_008761915.1 | 3 | 1493 | |||
| XM_008763694.1 | XP_008761916.1 | 4 | 1561 | |||
| GenBank mRNA | AJ302650.2 | - | 3 | 1417 | ||
| BC078739.1 | - | 4 | 1517 | 7111835 | ||
| FQ231115.1 | - | 1 | 165 | |||
| FQ232586.1 | - | 1 | 192 | |||

Hemgn
Genomic Map